Methyl 3-(cyanomethyl)-1H-indole-5-carboxylate 97+%

Supplier: Ambeed
Warning

A139005-100MG A139005-1G A139005-250MG
77149-976EA 28.84 USD
77149-976 77149-978 77149-980
Methyl 3-(cyanomethyl)-1H-indole-5-carboxylate 97+%
Methyl 3-(cyanomethyl)-1H-indole-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₂O₂
MW: 214.22 g/mol
MDL Number: MFCD09954797
CAS Number: 113438-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR