1H-Pyrrolo[2,3-b]pyridin-4-amine hydrochloride 97%

Supplier: Ambeed
Warning

A138240-1G A138240-250MG
77149-950EA 86.52 USD
77149-950 77149-952
1H-Pyrrolo[2,3-b]pyridin-4-amine hydrochloride 97%
1H-Pyrrolo[2,3-b]pyridin-4-amine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₇N₃·ClH
MW: 169.612 g/mol
MDL Number: MFCD08689965
CAS Number: 1134307-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR