1-(4-(2-(Pyrrolidin-1-yl)ethoxy)phenyl)guanidine 2,2,2-trifluoroacetate 97%
|
Warning
|
A138196-100MG
A138196-1G
A138196-250MG
77149-944
77149-946
77149-948
1-(4-(2-(Pyrrolidin-1-yl)ethoxy)phenyl)guanidine 2,2,2-trifluoroacetate 97%
1-(4-(2-(Pyrrolidin-1-yl)ethoxy)phenyl)guanidine 2,2,2-trifluoroacetate
Precaución: for Research Use Only
Formula:
C₁₅H₂₁F₃N₄O₃ MW: 362.35 g/mol |
MDL Number:
MFCD28986189 CAS Number: 1142945-81-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...