2-(2-Bromopyridin-4-yl)propan-2-ol 95%

Supplier: Ambeed
Warning

A137764-100MG A137764-1G A137764-250MG
77149-930EA 80 USD
77149-930 77149-932 77149-934
2-(2-Bromopyridin-4-yl)propan-2-ol 95%
2-(2-Bromopyridin-4-yl)propan-2-ol

Precaución: for Research Use Only

Formula: C₈H₁₀BrNO
MDL Number: MFCD20482555
CAS Number: 1055073-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR