TGR-1202 98+%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(1-(4-amino-3-(3-fluoro-4-isopropoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl)ethyl)-6-fluoro-3-(3-fluorophenyl)-4H-chromen-4-one, Umbralisib, RP5264

A137762-10MG A137762-1MG A137762-25MG A137762-2MG A137762-5MG A137762-50MG
77149-920EA 92.7 USD
77149-920 77149-922 77149-924 77149-926 77149-928 77685-184
TGR-1202 98+%
TGR-1202

Precaución: for Research Use Only

Formula: C₃₁H₂₄F₃NO₃
MW: 571.56 g/mol
MDL Number: MFCD28386165
CAS Number: 1532533-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR