cis-3-Hexenyl acetate 98%

Supplier: Ambeed
Danger

A137723-25G A137723-5G
77149-916EA 22.66 USD
77149-916 77149-918
cis-3-Hexenyl acetate 98%
cis-3-Hexenyl acetate

Precaución: for Research Use Only

Formula: C₈H₁₄O₂
MW: 142.20 g/mol
MDL Number: MFCD00036563
CAS Number: 3681-71-8
UN: 3272
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR