1-Methyl-1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A137483-1G A137483-250MG A137483-5G
77149-906EA 35 USD
77149-906 77149-908 77149-910
1-Methyl-1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid 95%
1-Methyl-1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MDL Number: MFCD15529044
CAS Number: 171919-37-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR