Fmoc-Trp(ME)-OH 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(1-methyl-1H-indol-3-yl)propanoic acid

A136921-100MG A136921-1G A136921-250MG A136921-5G
77149-888EA 22.66 USD
77149-888 77149-890 77149-892 77149-894
Fmoc-Trp(ME)-OH 98%
Fmoc-Trp(ME)-OH

Precaución: for Research Use Only

Formula: C₂₇H₂₄N₂O₄
MW: 440.499 g/mol
MDL Number: MFCD02684512
CAS Number: 1334509-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR