SCR7 pyrazine 95%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dihydro-6,7-diphenyl-2-thioxo-4(1H)-pteridinone, 6,7-Diphenyl-2-thioxo-2,3-dihydropteridin-4(1H)-one

A136439-10MG A136439-1MG A136439-25MG A136439-50MG A136439-5MG
77149-862EA 23 USD
77149-862 77149-864 77149-866 77149-868 77149-870
SCR7 pyrazine 95%
SCR7 pyrazine

Precaución: for Research Use Only

Formula: C₁₈H₁₂N₄OS
MDL Number: MFCD02167478
CAS Number: 14892-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR