GSK343 98%

Supplier: Ambeed
Warning

Synonyms: 1-Isopropyl-N-((6-methyl-2-oxo-4-propyl-1,2-dihydropyridin-3-yl)methyl)-6-(2-(4-methylpiperazin-1-yl)pyridin-4-yl)-1H-indazole-4-carboxamide

A135977-10MG A135977-1MG A135977-25MG A135977-5MG
77149-846EA 97.85 USD
77149-846 77149-848 77149-850 77149-852
GSK343 98%
GSK343

Precaución: for Research Use Only

Formula: C₃₁H₃₉N₇O₂
MW: 541.7 g/mol
MDL Number: MFCD25563250
CAS Number: 1346704-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR