Boc-Dab(Fmoc)-OH 97%

Supplier: Ambeed
Warning

A135795-10G A135795-1G A135795-250MG A135795-25G A135795-5G
77149-828EA 46 USD
77149-828 77149-830 77149-832 77149-834 77901-086
Boc-Dab(Fmoc)-OH 97%
Boc-Dab(Fmoc)-OH

Precaución: for Research Use Only

Formula: C₂₄H₂₈N₂O₆
MW: 440.496 g/mol
MDL Number: MFCD00236845
CAS Number: 117106-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR