Fmoc-α-Me-Trp(Boc)-OH 95%

Supplier: Ambeed
Warning

A135728-100MG A135728-1G A135728-250MG
77149-822EA 438.78 USD
77149-822 77149-824 77149-826
Fmoc-α-Me-Trp(Boc)-OH 95%
Fmoc-α-Me-Trp(Boc)-OH

Precaución: for Research Use Only

Formula: C₃₂H₃₂N₂O₆
MW: 540.60 g/mol
MDL Number: MFCD16652328
CAS Number: 1315449-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR