3-(3-Methoxyphenyl)-1-propene 97%

Supplier: Ambeed
Warning

A135492-250MG A135492-1G A135492-5G
77149-804EA 25 USD
77149-804 77902-018 77900-796
3-(3-Methoxyphenyl)-1-propene 97%
3-(3-Methoxyphenyl)-1-propene

Precaución: for Research Use Only

Formula: C₁₀H₁₂O
MDL Number: MFCD06201195
CAS Number: 24743-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR