(S,E)-2-(5-(3,7-Dimethylocta-2,6-dien-1-yl)-2,4-dihydroxyphenyl)-5,7-dihydroxychroman-4-one 95%

Supplier: Ambeed
Warning

A135368-1MG A135368-2MG A135368-5MG
77149-798EA 77.25 USD
77149-798 77149-800 77149-802
(S,E)-2-(5-(3,7-Dimethylocta-2,6-dien-1-yl)-2,4-dihydroxyphenyl)-5,7-dihydroxychroman-4-one 95%
(S,E)-2-(5-(3,7-Dimethylocta-2,6-dien-1-yl)-2,4-dihydroxyphenyl)-5,7-dihydroxychroman-4-one

Precaución: for Research Use Only

Formula: C₂₅H₂₈O₆
MW: 424.49 g/mol
MDL Number: MFCD21333736
CAS Number: 68401-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR