4,4,5,5-Tetramethyl-2-(2,4,6-trimethylphenyl)-1,3,2-dioxaborolane 95%

Supplier: Ambeed
Warning

A135325-1G A135325-25G A135325-5G
77149-794EA 20 USD
77149-794 77149-796 77566-650
4,4,5,5-Tetramethyl-2-(2,4,6-trimethylphenyl)-1,3,2-dioxaborolane 95%
4,4,5,5-Tetramethyl-2-(2,4,6-trimethylphenyl)-1,3,2-dioxaborolane

Precaución: For research use only.

Formula: C₁₅H₂₃BO₂
MDL Number: MFCD22414456
CAS Number: 171364-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR