2-(Methylsulfonyl)-2,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole benzenesulfonate 98%

Supplier: Ambeed
Warning

A134910-1G A134910-250MG A134910-5G
77149-786EA 28.84 USD
77149-786 77149-788 77149-790
2-(Methylsulfonyl)-2,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole benzenesulfonate 98%
2-(Methylsulfonyl)-2,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole benzenesulfonate

Precaución: for Research Use Only

Formula: C₁₂H₁₅N₃O₅S₂
MW: 345.39 g/mol
MDL Number: MFCD28144046
CAS Number: 1280210-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR