3-Aminobenzofuran-2-carboxamide 95%

Supplier: Ambeed
Warning

A134501-1G A134501-250MG
77149-778EA 19.57 USD
77149-778 77149-780
3-Aminobenzofuran-2-carboxamide 95%
3-Aminobenzofuran-2-carboxamide

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MW: 176.18 g/mol
MDL Number: MFCD00457147
CAS Number: 54802-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR