Methyl 5-bromo-7-azaindole-6-carboxylate 97%

Supplier: Ambeed
Warning

A134144-100MG A134144-250MG A134144-1G
77149-766EA 27 USD
77149-766 77149-768 77566-628
Methyl 5-bromo-7-azaindole-6-carboxylate 97%
Methyl 5-bromo-7-azaindole-6-carboxylate

Precaución: For research use only.

Formula: C₉H₇BrN₂O₂
MDL Number: MFCD21606695
CAS Number: 1408074-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR