1-Chloro-2-fluoro-5-iodobenzene 98%
Supplier: Ambeed
|
Warning
|
A133752-100G
A133752-10G
A133752-25G
A133752-5G
77149-726EA
58
USD
77149-726
77149-728
77149-730
77149-732
1-Chloro-2-fluoro-5-iodobenzene 98%
1-Chloro-2-fluoro-5-iodobenzene
Precaución: for Research Use Only
Formula:
C₆H₃ClFI |
MDL Number:
MFCD00042210 CAS Number: 156150-67-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...