O-[(Ethoxycarbonyl)cyanomethylenamino]-N,N,N',N'-tetramethyluronium Hexafluorophosphate 98%

Supplier: Ambeed
Warning

Synonyms: HOTU, N-(6-Cyano-2-methyl-7-oxo-4,8-dioxa-2,5-diazadec-5-en-3-ylidene)-N-methylmethanaminium hexafluorophosphate(V)

A133485-10G A133485-25G A133485-5G
77149-720EA 38.11 USD
77149-720 77149-722 77149-724
O-[(Ethoxycarbonyl)cyanomethylenamino]-N,N,N',N'-tetramethyluronium Hexafluorophosphate 98%
O-[(Ethoxycarbonyl)cyanomethylenamino]-N,N,N',N'-tetramethyluronium Hexafluorophosphate

Precaución: for Research Use Only

Formula: C₁₀H₁₇F₆N₄O₃P
MW: 386.24 g/mol
MDL Number: MFCD01862832
CAS Number: 333717-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR