endo-2-(Boc-amino)-8-azabicyclo[3.2.1]octane 95%

Supplier: Ambeed
Warning

A133244-100MG A133244-50MG
77149-716EA 458.35 USD
77149-716 77149-718
endo-2-(Boc-amino)-8-azabicyclo[3.2.1]octane 95%
endo-2-(Boc-amino)-8-azabicyclo[3.2.1]octane

Precaución: for Research Use Only

Formula: C₁₂H₂₂N₂O₂
MW: 226.31 g/mol
MDL Number: MFCD23381650
CAS Number: 1408076-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR