4-Fluoro-3-(trifluoromethoxy)phenol 98%

Supplier: Ambeed
Warning

A133173-100MG A133173-1G A133173-250MG
77149-710EA 53 USD
77149-710 77149-712 77149-714
4-Fluoro-3-(trifluoromethoxy)phenol 98%
4-Fluoro-3-(trifluoromethoxy)phenol

Precaución: for Research Use Only

Formula: C₇H₄F₄O₂
MDL Number: MFCD06660257
CAS Number: 886501-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR