Golvatinib 99+%

Supplier: Ambeed
Warning

Synonyms: E7050, E 7050, N-(2-fluoro-4-((2-(4-(4-methylpiperazin-1-yl)piperidine-1-carboxamido)pyridin-4-yl)oxy)phenyl)-N-(4-fluorophenyl)cyclopropane-1,1-dicarboxamide, E-7050

A133170-10MG A133170-1MG A133170-25MG A133170-5MG
77149-702EA 59.74 USD
77149-702 77149-704 77149-706 77149-708
Golvatinib 99+%
Golvatinib

Precaución: for Research Use Only

Formula: C₃₃H₃₇F₂N₇O₄
MW: 633.69 g/mol
MDL Number: MFCD22124462
CAS Number: 928037-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR