Methyl (3R)-3-aminobutanoate hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-3-Amino-butyric acid methyl ester hydrochloride, (R)-Methyl 3-aminobutanoate hydrochloride
A133136-10G
A133136-1G
A133136-25G
A133136-5G
77149-694EA
60.77
USD
77149-694
77149-696
77149-698
77149-700
Methyl (3R)-3-aminobutanoate hydrochloride 97%
Methyl (3R)-3-aminobutanoate hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₁NO₂·ClH MW: 153.607 g/mol |
MDL Number:
MFCD12068616 CAS Number: 139243-54-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...