1-(2,7-Dibromospiro[fluorene-9,4'-piperidin]-1'-yl)ethanone 97%

Supplier: Ambeed
Warning

A133096-100MG A133096-1G A133096-250MG
77149-688EA 16.48 USD
77149-688 77149-690 77149-692
1-(2,7-Dibromospiro[fluorene-9,4'-piperidin]-1'-yl)ethanone 97%
1-(2,7-Dibromospiro[fluorene-9,4'-piperidin]-1'-yl)ethanone

Precaución: for Research Use Only

Formula: C₁₉H₁₇Br₂NO
MW: 435.15 g/mol
MDL Number: MFCD28144724
CAS Number: 1616114-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR