MC1568 96%

Supplier: Ambeed
Warning

Synonyms: (E)-3-(5-((E)-3-(3-Fluorophenyl)-3-oxoprop-1-en-1-yl)-1-methyl-1H-pyrrol-2-yl)-N-hydroxyacrylamide, 3-[5-(3-(3-Fluorophenyl)-3-oxopropen-1-yl)-1-methyl-1H-pyrrol-2-yl]-N-hydroxy-2-propenamide, MC-1568

A133036-10MG A133036-1MG A133036-50MG
77149-682EA 46.35 USD
77149-682 77149-684 77149-686
MC1568 96%
MC1568

Precaución: for Research Use Only

Formula: C₁₇H₁₅FN₂O₃
MW: 314.31 g/mol
MDL Number: MFCD16875423
CAS Number: 852475-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR