3-Bromo-4-methoxy-5-nitro-benzoic acid 95%

Supplier: Ambeed
Warning

A132986-100MG A132986-1G A132986-250MG A132986-5G
77149-674EA 30.9 USD
77149-674 77149-676 77149-678 77149-680
3-Bromo-4-methoxy-5-nitro-benzoic acid 95%
3-Bromo-4-methoxy-5-nitro-benzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrNO₅
MW: 276.04 g/mol
MDL Number: MFCD09881755
CAS Number: 954815-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR