Benzyl 3-((tert-butoxycarbonyl)amino)-5-hydroxypiperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A132777-100MG A132777-1G A132777-250MG
77149-668EA 155 USD
77149-668 77149-670 77149-672
Benzyl 3-((tert-butoxycarbonyl)amino)-5-hydroxypiperidine-1-carboxylate 95%
Benzyl 3-((tert-butoxycarbonyl)amino)-5-hydroxypiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₈H₂₆N₂O₅
MW: 350.40 g/mol
MDL Number: MFCD26727374
CAS Number: 1785642-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR