H-Pro-NHEt·HCl 97%

Supplier: Ambeed
Warning

Synonyms: H-Pro-NHEt·HCl, (S)-N-Ethylpyrrolidine-2-carboxamide hydrochloride

A132289-10G A132289-25G
77149-664EA 64.89 USD
77149-664 77611-176
H-Pro-NHEt·HCl 97%
H-Pro-NHEt·HCl

Precaución: for Research Use Only

Formula: C₇H₁₅ClN₂O
MW: 178.66 g/mol
MDL Number: MFCD00238159
CAS Number: 58107-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR