4-Chlorobenzo[b]thiophene 97%

Supplier: Ambeed
Warning

A131858-100G A131858-10G A131858-1G A131858-25G A131858-5G
77149-644EA 107 USD
77149-644 77149-646 77149-648 77149-650 77149-652
4-Chlorobenzo[b]thiophene 97%
4-Chlorobenzo[b]thiophene

Precaución: for Research Use Only

Formula: C₈H₅ClS
MDL Number: MFCD18451627
CAS Number: 66490-33-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR