2-Azaspiro[3.3]heptane hydrochloride 98%

Supplier: Ambeed
Warning

A131624-250MG A131624-5G A131624-1G A131624-100MG A131624-25G
77149-636EA 27 USD
77149-636 77149-638 77549-566 77891-921 77891-922
2-Azaspiro[3.3]heptane hydrochloride 98%
2-Azaspiro[3.3]heptane hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂ClN
MDL Number: MFCD21607345
CAS Number: 1420271-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR