N'-(3-Allyl-2-hydroxybenzylidene)-2-(4-benzylpiperazin-1-yl)acetohydrazide 98%

Supplier: Ambeed
Danger

Synonyms: PAC-1

A131202-10MG A131202-1MG A131202-50MG A131202-5MG A131202-250MG A131202-100MG
77149-616EA 12.36 USD
77149-616 77149-618 77149-620 77149-622 77566-546 77685-140
N'-(3-Allyl-2-hydroxybenzylidene)-2-(4-benzylpiperazin-1-yl)acetohydrazide 98%
N'-(3-Allyl-2-hydroxybenzylidene)-2-(4-benzylpiperazin-1-yl)acetohydrazide

Precaución: For research use only.

Formula: C₂₃H₂₈N₄O₂
MW: 392.49 g/mol
MDL Number: MFCD02051977
CAS Number: 315183-21-2
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR