Pyrazine-2-carboximidamide hydrochloride 97%

Supplier: Ambeed
Warning

A130843-25G A130843-5G A130843-1G
77149-612EA 101.97 USD
77149-612 77149-614 77685-138
Pyrazine-2-carboximidamide hydrochloride 97%
Pyrazine-2-carboximidamide hydrochloride

Precaución: for Research Use Only

Formula: C5H7ClN4
MW: 158.5900 g/mol
MDL Number: MFCD05663360
CAS Number: 138588-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR