WIKI-4 98%

Supplier: Ambeed
Warning

Synonyms: 2-(3-((4-(4-Methoxyphenyl)-5-(pyridin-4-yl)-4H-1,2,4-triazol-3-yl)thio)propyl)-1H-benzo[de]isoquinoline-1,3(2H)-dione, WIKI-4, WIKI 4, 1-[(1S)-1-(4-chloro-3-fluorophenyl)-2-hydroxyethyl]-4-[2-[(2-methylpyrazol-3-yl)amino]pyrimidin-4-yl]pyridin-2-one, WIKI4

A130685-10MG A130685-1MG A130685-25MG A130685-50MG A130685-5MG
77149-590EA 157.59 USD
77149-590 77149-592 77149-594 77149-596 77149-598
WIKI-4 98%
WIKI-4

Precaución: for Research Use Only

Formula: C₂₉H₂₃N₅O₃S
MW: 521.59 g/mol
MDL Number: MFCD05885187
CAS Number: 838818-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR