(1S,3S,5S)-2-((2S)-2-Amino-2-(3-hydroxyadamantan-1-yl)acetyl)-2-azabicyclo[3.1.0]hexane-3-carbonitrile hydrochloride 98%

Supplier: Ambeed
Warning

A130556-100MG A130556-1MG
77149-578EA 23.69 USD
77149-578 77149-580
(1S,3S,5S)-2-((2S)-2-Amino-2-(3-hydroxyadamantan-1-yl)acetyl)-2-azabicyclo[3.1.0]hexane-3-carbonitrile hydrochloride 98%
(1S,3S,5S)-2-((2S)-2-Amino-2-(3-hydroxyadamantan-1-yl)acetyl)-2-azabicyclo[3.1.0]hexane-3-carbonitrile hydrochloride

Precaución: for Research Use Only

Formula: C₁₈H₂₆ClN₃O₂
MW: 351.87 g/mol
MDL Number: MFCD22572334
CAS Number: 709031-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR