Calcium pantothenate 95%

Supplier: Ambeed
Warning

A130216-100G A130216-10G A130216-25G A130216-500G A130216-5G A130216-1000G
77149-552EA 18 USD
77149-552 77149-554 77149-556 77149-558 77149-560 77685-130
Calcium pantothenate 95%
Calcium pantothenate

Precaución: for Research Use Only

Formula: C₁₈H₃₂CaN₂O₁₀
MDL Number: MFCD00002766
CAS Number: 137-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR