Lomeguatrib 98%

Supplier: Ambeed
Warning

Synonyms: 6-((4-Bromothiophen-2-yl)methoxy)-9H-purin-2-amine

A129482-10MG A129482-1MG A129482-50MG A129482-5MG
77149-544EA 167.89 USD
77149-544 77149-546 77149-548 77149-550
Lomeguatrib 98%
Lomeguatrib

Precaución: for Research Use Only

Formula: C₁₀H₈BrN₅OS
MW: 326.1700 g/mol
MDL Number: MFCD23703756
CAS Number: 192441-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR