7-Bromo-6-fluoro-2,3-dihydro-1H-inden-1-one 95%

Supplier: Ambeed
Warning

A129206-100MG A129206-1G A129206-250MG
77149-528EA 91.67 USD
77149-528 77149-530 77149-532
7-Bromo-6-fluoro-2,3-dihydro-1H-inden-1-one 95%
7-Bromo-6-fluoro-2,3-dihydro-1H-inden-1-one

Precaución: for Research Use Only

Formula: C₉H₆BrFO
MW: 229.05 g/mol
MDL Number: MFCD18208172
CAS Number: 1260011-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR