Methyl (3S,4S)-1-benzyl-4-methylpyrrolidine-3-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: (3S,4S)-Methyl 1-benzyl-4-methylpyrrolidine-3-carboxylate, (3S,4S)-1-Benzyl-3-(methoxycarbonyl)-4-methylpyrrolidine

A129013-100MG A129013-1G A129013-250MG
77149-522EA 23.69 USD
77149-522 77149-524 77149-526
Methyl (3S,4S)-1-benzyl-4-methylpyrrolidine-3-carboxylate 95%
Methyl (3S,4S)-1-benzyl-4-methylpyrrolidine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₂
MW: 233.306 g/mol
MDL Number: MFCD22420558
CAS Number: 181114-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR