trans-(4-(Trifluoromethyl)cyclohexyl)methanol 97%

Supplier: Ambeed
Warning

A128975-1G A128975-250MG A128975-100MG
77149-520EA 81 USD
77149-520 77611-140 77900-730
trans-(4-(Trifluoromethyl)cyclohexyl)methanol 97%
trans-(4-(Trifluoromethyl)cyclohexyl)methanol

Precaución: for Research Use Only

Formula: C₈H₁₃F₃O
MDL Number: MFCD29054589
CAS Number: 1202577-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR