4-Bromo-2-fluoro-6-nitrophenol 98%

Supplier: Ambeed
Danger

A128312-100G A128312-10G A128312-1G A128312-250MG A128312-25G A128312-500G A128312-5G
77149-492EA 260 USD
77149-492 77149-494 77149-496 77149-498 77149-500 77149-502 77149-504
4-Bromo-2-fluoro-6-nitrophenol 98%
4-Bromo-2-fluoro-6-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₃BrFNO₃
MDL Number: MFCD00013372
CAS Number: 320-76-3
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR