CC-122 95%

Supplier: Ambeed
Warning

A128261-250MG A128261-100MG A128261-1G A128261-50MG
77149-486EA 9.27 USD
77149-486 77149-488 77149-490 77685-112 77901-253 77901-417 77901-767
CC-122 95%
CC-122

Precaución: for Research Use Only

Formula: C₁₄H₁₄N₄O₃
MDL Number: MFCD29919294
CAS Number: 1015474-32-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR