BMS-626529 98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-benzoylpiperazin-1-yl)-2-(4-methoxy-7-(3-methyl-1H-1,2,4-triazol-1-yl)-1H-pyrrolo[2,3-c]pyridin-3-yl)ethane-1,2-dione, Temsavir

A128252-10MG A128252-1MG A128252-5MG A128252-50MG
77149-480EA 22.66 USD
77149-480 77149-482 77149-484 77685-111
BMS-626529 98%
BMS-626529

Precaución: for Research Use Only

Formula: C₂₄H₂₃NO₄
MW: 473.48 g/mol
MDL Number: MFCD22665723
CAS Number: 701213-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR