Fmoc-(Dmb)Gly-OH 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-(Dmb)Gly-OH

A127852-100MG A127852-1G A127852-250MG A127852-5G A127852-25G
77149-470EA 7 USD
77149-470 77149-472 77149-474 77549-520 77891-897
Fmoc-(Dmb)Gly-OH 98%
Fmoc-(Dmb)Gly-OH

Precaución: for Research Use Only

Formula: C₂₆H₂₅NO₆
MW: 447.487 g/mol
MDL Number: MFCD08064316
CAS Number: 166881-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR