Dichlorobis(di-tert-butylphenylphosphine)palladium(II) 97%

Supplier: Ambeed
Warning

Synonyms: Dichlorobis(di-tert-butylphenylphosphinE)palladium(ii)

A126344-1G A126344-250MG A126344-5G
77149-434EA 35 USD
77149-434 77149-436 77901-064
Dichlorobis(di-tert-butylphenylphosphine)palladium(II) 97%
Dichlorobis(di-tert-butylphenylphosphine)palladium(II)

Precaución: for Research Use Only

Formula: C₂₈H₄₆Cl₂P₂Pd
MW: 621.944 g/mol
MDL Number: MFCD09953450
CAS Number: 34409-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR