Sarcosine 98%

Supplier: Ambeed
Warning

Synonyms: H-Sar-OH, N-Methylglycine, Methylaminoacetic acid

A125930-100G A125930-25MG A125930-25G A125930-1000G A125930-500G A125930-100MG
77149-430EA 19.57 USD
77149-430 77149-432 77312-902 77611-076 77611-078 77685-101
Sarcosine 98%
Sarcosine

Precaución: for Research Use Only

Formula: C₃H₇NO₂
MW: 89.09 g/mol
MDL Number: MFCD00004279
CAS Number: 107-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR