(3R,4S)-rel-tert-Butyl 3-hydroxy-4-methylpyrrolidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A125768-250MG A125768-1G
77149-428EA 169.95 USD
77149-428 77685-097
(3R,4S)-rel-tert-Butyl 3-hydroxy-4-methylpyrrolidine-1-carboxylate 97%
(3R,4S)-rel-tert-Butyl 3-hydroxy-4-methylpyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₃
MW: 201.26 g/mol
MDL Number: MFCD18375158
CAS Number: 885102-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR