2-(Boc-amino)-4-methylthiophene 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (4-methylthiophen-2-yl)carbamate

A125709-1G A125709-250MG A125709-5G
77149-418EA 17.51 USD
77149-418 77149-420 77149-422
2-(Boc-amino)-4-methylthiophene 97%
2-(Boc-amino)-4-methylthiophene

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO₂S
MW: 213.3 g/mol
MDL Number: MFCD21642065
CAS Number: 887475-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR