Ethyl 2-bromo-4,5-difluorobenzoate 95%
Supplier: Ambeed
|
Warning
|
A125242-1G
A125242-25G
A125242-5G
77149-406EA
7.21
USD
77149-406
77149-408
77149-410
Ethyl 2-bromo-4,5-difluorobenzoate 95%
Ethyl 2-bromo-4,5-difluorobenzoate
Precaución: for Research Use Only
Formula:
C₉H₇BrF₂O₂ MW: 265.051 g/mol |
MDL Number:
MFCD08458105 CAS Number: 144267-97-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...