1-Cbz-3-Cyanopiperidine 97%
Supplier: Ambeed
|
Warning
|
A125224-100G
A125224-10G
A125224-1G
A125224-5G
A125224-25G
77149-398EA
615.94
USD
77149-398
77149-400
77149-402
77149-404
77683-114
1-Cbz-3-Cyanopiperidine 97%
1-Cbz-3-Cyanopiperidine
Precaución: For research use only.
Formula:
C₁₄H₁₆N₂O₂ MW: 244.29 g/mol |
MDL Number:
MFCD04115339 CAS Number: 885069-22-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...